Methyl-N-{[(2-methyl-2-propanyl)oxy]carbonyl}glycinat 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD33192-500GEA 304 EUR
BLDPBD33192-500G BLDPBD33192-10G BLDPBD33192-25G BLDPBD33192-5G BLDPBD33192-100G
Methyl-N-{[(2-methyl-2-propanyl)oxy]carbonyl}glycinat 97%
Boc-glycine methyl ester

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₈H₁₅NO₄
Molekulargewicht: 189,2 g/mol
Lagerungstemperatur: Raumtemperatur
CAS-Nummer: 31954-27-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR