5-Bromnicotinamid 97%

BLDPBD3334-100GEA 150 EUR
BLDPBD3334-100G BLDPBD3334-1G BLDPBD3334-25G BLDPBD3334-5G BLDPBD3334-10G
5-Bromnicotinamid 97%
5-Bromnicotinamid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₅BrN₂O
Molekulargewicht: 201,03 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00173919
CAS-Nummer: 28733-43-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR