Ethyl-6-fluorimidazo[1,2-a]pyridin-2-carboxylat 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD33417-100MGEA 18 EUR
BLDPBD33417-100MG BLDPBD33417-250MG
Ethyl-6-fluorimidazo[1,2-a]pyridin-2-carboxylat 97%
Ethyl-6-fluorimidazo[1,2-a]pyridin-2-carboxylat

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₉FN₂O₂
Molekulargewicht: 208,1900 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06659035
CAS-Nummer: 367500-93-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR