6-Chloro-5-cyanonicotinic acid 95%

BLDPBD337829-250MGEA 146 EUR
BLDPBD337829-250MG BLDPBD337829-1G BLDPBD337829-100MG
6-Chloro-5-cyanonicotinic acid 95%
6-Chloro-5-cyanonicotinic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₃ClN₂O₂
Molekulargewicht: 182,564 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD14585457
CAS-Nummer: 1206969-20-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR