4-Fluor-3-iod-1H-indazol 97%
|
|
BLDPBD33903-250MGEA
19
EUR
BLDPBD33903-250MG
4-Fluor-3-iod-1H-indazol 97%
4-Fluoro-3-iodo-1H-indazole
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formel:
C₇H₄FIN₂ Molekulargewicht: 262,0200 g/mol Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD07781590 CAS-Nummer: 518990-32-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...