Hesperetin 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD33910-5GEA 9 EUR
BLDPBD33910-5G BLDPBD33910-10G BLDPBD33910-500G BLDPBD33910-100G BLDPBD33910-25G BLDPBD33910-1G
Hesperetin 97%
Hesperetin

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₆H₁₄O₆
Molekulargewicht: 302,30 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00075646
CAS-Nummer: 520-33-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR