5-Amino-2-pyridincarbonitril 96%

Lieferant: BLD PHARMATECH GMBH

BLDPBD3408-25GEA 542 EUR
BLDPBD3408-25G BLDPBD3408-1G BLDPBD3408-100MG BLDPBD3408-5G BLDPBD3408-250MG
5-Amino-2-pyridincarbonitril 96%
5-Amino-2-pyridincarbonitril

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₆H₅N₃
Molekulargewicht: 119,1240 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03093956
CAS-Nummer: 55338-73-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR