Imidazo[1,2-a]pyridin-3-carbaldehyd 98%

BLDPBD34329-25GEA 745 EUR
BLDPBD34329-25G BLDPBD34329-5G BLDPBD34329-1G BLDPBD34329-100MG BLDPBD34329-250MG
Imidazo[1,2-a]pyridin-3-carbaldehyd 98%
Imidazo[1,2-a]pyridin-3-carbaldehyd

Storage condition: Inert atmosphere, 2-8°C

Formel: C₈H₆N₂O
Molekulargewicht: 146,1500 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD07778354
CAS-Nummer: 6188-43-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR