DL-Ethionin 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD34574-1GEA 45 EUR
BLDPBD34574-1G BLDPBD34574-25G BLDPBD34574-5G
DL-Ethionin 98%
DL-Ethionin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₆H₁₃NO₂S
Molekulargewicht: 163,24 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00063102
CAS-Nummer: 67-21-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR