tert-Butylhydrazin Hydrochlorid 95%
|
|
BLDPBD34793-100GEA
13
EUR
BLDPBD34793-100G
BLDPBD34793-10G
BLDPBD34793-1G
BLDPBD34793-1000G
BLDPBD34793-0.025G
BLDPBD34793-500G
BLDPBD34793-25G
tert-Butylhydrazin Hydrochlorid 95%
tert-Butylhydrazin Hydrochlorid
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formel:
C₄H₁₃ClN₂ Molekulargewicht: 124,61 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00012947 CAS-Nummer: 7400-27-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...