5-Nitro-2-picolin 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD3510-10GEA 27 EUR
BLDPBD3510-10G BLDPBD3510-100G BLDPBD3510-25G BLDPBD3510-5G BLDPBD3510-1G
5-Nitro-2-picolin 97%
5-Nitro-2-picolin

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₆N₂O₂
Molekulargewicht: 138,12 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD04114179
CAS-Nummer: 21203-68-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR