3-(Morpholino)phenylboronsäurepinakolester 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD35121-100MGEA 11 EUR
BLDPBD35121-100MG BLDPBD35121-25G BLDPBD35121-250MG BLDPBD35121-5G BLDPBD35121-1G BLDPBD35121-10G
3-(Morpholino)phenylboronsäurepinakolester 98%
3-(Morpholino)phenylboronsäurepinakolester

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₆H₂₄BNO₃
Molekulargewicht: 289,1777 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03412097
CAS-Nummer: 852227-95-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR