2-Aminophenylboronsäure Hydrochlorid 97%

BLDPBD3663-25GEA 219 EUR
BLDPBD3663-25G BLDPBD3663-100G BLDPBD3663-5G BLDPBD3663-250MG BLDPBD3663-1G BLDPBD3663-10G
2-Aminophenylboronsäure Hydrochlorid 97%
2-Aminophenylboronsäure Hydrochlorid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₉BClNO₂
Molekulargewicht: 173,4052 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD02258096
CAS-Nummer: 863753-30-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR