2-Bromo-3-chloro-4-methylpyridine 98%

BLDPBD36992-5GEA 120 EUR
BLDPBD36992-5G BLDPBD36992-1G
2-Bromo-3-chloro-4-methylpyridine 98%
2-Bromo-3-chloro-4-methylpyridine

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₅BrClN
Molekulargewicht: 206,468 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD08277283
CAS-Nummer: 884495-42-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR