2-Chlor-5-fluorisonicotinsäure 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD37056-25GEA 381 EUR
BLDPBD37056-25G BLDPBD37056-100MG BLDPBD37056-1G BLDPBD37056-250MG BLDPBD37056-5G
2-Chlor-5-fluorisonicotinsäure 98%
2-Chlor-5-fluorisonicotinsäure

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₃ClFNO₂
Molekulargewicht: 175,5500 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD04972381
CAS-Nummer: 884494-74-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR