4-(2-Aminoethyl)tetrahydropyran 97%

BLDPBD3942-5GEA 279 EUR
BLDPBD3942-5G BLDPBD3942-1G BLDPBD3942-100MG BLDPBD3942-25G BLDPBD3942-250MG BLDPBD3942-10G
4-(2-Aminoethyl)tetrahydropyran 97%
4-(2-Aminoethyl)tetrahydropyran

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₇H₁₅NO
Molekulargewicht: 129,2001 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD02179433
CAS-Nummer: 65412-03-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR