6,7-Dimethoxycoumarin 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD39903-250MGEA 82 EUR
BLDPBD39903-250MG BLDPBD39903-1G BLDPBD39903-5G BLDPBD39903-100MG
6,7-Dimethoxycoumarin 95%
6,7-Dimethoxycoumarin

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₁H₁₀O₄
Molekulargewicht: 206,195 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00006871
CAS-Nummer: 120-08-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR