2,2'-Dipyridyldisulfid 98%

Lieferant: BLD PHARMATECH GMBH

Synonyme: 2,2'-Dithiodipyridin

BLDPBD41054-1000GEA 896 EUR
BLDPBD41054-1000G BLDPBD41054-25G BLDPBD41054-100G BLDPBD41054-5G BLDPBD41054-500G
2,2'-Dipyridyldisulfid 98%
2,2'-Dipyridyldisulfid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₀H₈N₂S₂
Molekulargewicht: 220,32 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00006287
CAS-Nummer: 2127-03-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR