2-Mercaptoethansulfonsäure Natriumsalz (Mesna) 95+%

BLDPBD41084-1GEA 9 EUR
BLDPBD41084-1G BLDPBD41084-25G BLDPBD41084-5G BLDPBD41084-10G
2-Mercaptoethansulfonsäure Natriumsalz (Mesna) 95+%
2-Mercaptoethansulfonsäure Natriumsalz (Mesna)
Formel: C₂H₅NaO₃S₂
Molekulargewicht: 164,18 g/mol
MDL Number: MFCD00007535
CAS-Nummer: 19767-45-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR