5,7-Dimethyl-8-chinolinol 97%
|
|
BLDPBD41643-1GEA
359
EUR
BLDPBD41643-1G
BLDPBD41643-250MG
BLDPBD41643-100MG
BLDPBD41643-5G
5,7-Dimethyl-8-chinolinol 97%
5,7-Dimethyl-8-chinolinol
Storage condition: Sealed in dry, Room Temperature
Formel:
C₁₁H₁₁NO Molekulargewicht: 173,2111 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00168976 CAS-Nummer: 37873-29-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...