L(+)-Histidin Hydrochlorid Monohydrat 98%
|
|
Synonyme:
H-His-OH.HCl.H2O
, His HCl Monohydrat
, (S)-(+)-Histidin Hydrochlorid Monohydrat
, L-3-Imidazol-4-ylalanin Hydrochlorid Monohydrat
, (S)-(+)-2-Amino-3-(4-imidazoyl)propansäure Hydrochlorid Monohydrat
, (2S)-(+)-Histidin Hydrochlorid Monohydrat
BLDPBD41834-500GEA
105
EUR
BLDPBD41834-500G
BLDPBD41834-25G
BLDPBD41834-100G
BLDPBD41834-1000G
L(+)-Histidin Hydrochlorid Monohydrat 98%
L(+)-Histidin Hydrochlorid Monohydrat
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₆H₉N₃O₂·HCl·H₂O Molekulargewicht: 209,63 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00151027 CAS-Nummer: 5934-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...