2-Hydroxy-3,5-dibromo-4-methylpyridine 95%

BLDPBD42090-5GEA 357 EUR
BLDPBD42090-5G BLDPBD42090-10G BLDPBD42090-250MG BLDPBD42090-25G BLDPBD42090-100MG BLDPBD42090-1G
2-Hydroxy-3,5-dibromo-4-methylpyridine 95%
2-Hydroxy-3,5-dibromo-4-methylpyridine

Storage condition: Sealed in dry, Room Temperature

Formel: C₆H₅Br₂NO
Molekulargewicht: 266,918 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06254604
CAS-Nummer: 89581-53-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR