Tetrabutylphosphoniumhexafluorophosphat 98%

BLDPBD433811-25GEA 94 EUR
BLDPBD433811-25G BLDPBD433811-100G BLDPBD433811-5G BLDPBD433811-1G
Tetrabutylphosphoniumhexafluorophosphat 98%
Tetrabutylphosphonium hexafluorophosphate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₆H₃₆F₆P₂
Molekulargewicht: 404,3950 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01863114
CAS-Nummer: 111928-21-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR