m-PEG5-amine 98+%

BLDPBD434599-5GEA 645 EUR
BLDPBD434599-5G BLDPBD434599-250MG BLDPBD434599-1G BLDPBD434599-100MG
m-PEG5-amine 98+%
m-PEG5-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₁H₂₅NO₅
Molekulargewicht: 251,32 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD27977523
CAS-Nummer: 5498-83-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR