(4R)-2,2-Dioxido-4- phenyl -1,2,3-oxathiazolidine, N-BOC protected 98%
|
|
BLDPBD447679-100MGEA
58
EUR
BLDPBD447679-100MG
BLDPBD447679-250MG
BLDPBD447679-1G
(4R)-2,2-Dioxido-4- phenyl -1,2,3-oxathiazolidine, N-BOC protected 98%
(4R)-2,2-Dioxido-4- phenyl -1,2,3-oxathiazolidine, N-BOC protected
Storage condition: Sealed in dry, 2-8°C
Formel:
C₁₃H₁₇NO₅S Molekulargewicht: 299,343 g/mol Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD17018793 CAS-Nummer: 1209467-60-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...