(3-Chlorpropyl)trimethylsilan 98%

BLDPBD45103-25GEA 76 EUR
BLDPBD45103-0.025G BLDPBD45103-25G BLDPBD45103-5G
(3-Chlorpropyl)trimethylsilan 98%
(3-Chloropropyl)trimethylsilane

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₁₅ClSi
Molekulargewicht: 150,73 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00045299
CAS-Nummer: 2344-83-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR