2-Brom-3-nitrotoluol 98%
|
|
BLDPBD4526-100GEA
159
EUR
BLDPBD4526-100G
BLDPBD4526-25G
BLDPBD4526-10G
BLDPBD4526-5G
BLDPBD4526-1G
2-Brom-3-nitrotoluol 98%
2-Brom-3-nitrotoluol
Storage condition: Sealed in dry, Room Temperature
Formel:
C₇H₆BrNO₂ Molekulargewicht: 216,0300 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00134555 CAS-Nummer: 41085-43-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...