4-Boronobenzolsulfonamid 98%

BLDPBD45490-25GEA 969 EUR
BLDPBD45490-25G BLDPBD45490-10G BLDPBD45490-1G BLDPBD45490-250MG BLDPBD45490-5G BLDPBD45490-100MG
4-Boronobenzolsulfonamid 98%
4-Boronobenzolsulfonamid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₈BNO₄S
Molekulargewicht: 201,00 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD06659839
CAS-Nummer: 613660-87-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR