2,6-Dichlor-m-toluidin 98%

BLDPBD4753-25GEA 33 EUR
BLDPBD4753-25G BLDPBD4753-100G BLDPBD4753-5G
2,6-Dichlor-m-toluidin 98%
2,6-Dichlor-m-toluidin

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₇Cl₂N
Molekulargewicht: 176,0432 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00007676
CAS-Nummer: 64063-37-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR