Methyl-3-methoxy-2-thiophencarboxylat 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD5022-10GEA 317 EUR
BLDPBD5022-10G BLDPBD5022-1G BLDPBD5022-250MG BLDPBD5022-5G
Methyl-3-methoxy-2-thiophencarboxylat 97%
Methyl-3-methoxy-2-thiophencarboxylat

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₈O₃S
Molekulargewicht: 172,20 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00102115
CAS-Nummer: 62353-75-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR