3-Thien-3-yl-L-alanine 97%

BLDPBD5091-1GEA 96 EUR
BLDPBD5091-1G BLDPBD5091-250MG BLDPBD5091-5G
3-Thien-3-yl-L-alanine 97%
3-Thien-3-yl-L-alanine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₇H₉NO₂S
Molekulargewicht: 171,217 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00038645
CAS-Nummer: 3685-51-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR