4-Amino-2,5-dimethylphenol 97%
|
|
BLDPBD5316-1GEA
22
EUR
BLDPBD5316-1G
BLDPBD5316-25G
BLDPBD5316-10G
BLDPBD5316-100G
BLDPBD5316-5G
BLDPBD5316-250MG
BLDPBD5316-0.025G
4-Amino-2,5-dimethylphenol 97%
4-Amino-2,5-xylenol
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₈H₁₁NO Molekulargewicht: 137,1800 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00007873 CAS-Nummer: 3096-71-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...