cis-1,4-Cyclohexandicarbonsäure 90%

BLDPBD5391-1GEA 13 EUR
BLDPBD5391-1G BLDPBD5391-25G BLDPBD5391-5G BLDPBD5391-10G BLDPBD5391-500G BLDPBD5391-250MG BLDPBD5391-100G
cis-1,4-Cyclohexandicarbonsäure 90%
cis-1,4-Cyclohexanedicarboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₁₂O₄
Molekulargewicht: 172,1800 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00064950
CAS-Nummer: 619-81-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR