1,3-Dimethyl-1H-pyrazol 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD54179-100GEA 119 EUR
BLDPBD54179-100G BLDPBD54179-10G BLDPBD54179-5G BLDPBD54179-500G BLDPBD54179-25G BLDPBD54179-1G
1,3-Dimethyl-1H-pyrazol 98%
1,3-Dimethyl-1H-pyrazole

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₅H₈N₂
Molekulargewicht: 96,1304 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00036665
CAS-Nummer: 694-48-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR