1,3-Di-o-tolylguanidin 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD54209-100GEA 34 EUR
BLDPBD54209-100G BLDPBD54209-500G BLDPBD54209-25G
1,3-Di-o-tolylguanidin 98%
1,3-Di-o-tolylguanidin

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₅H₁₇N₃
Molekulargewicht: 239,32 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00008513
CAS-Nummer: 97-39-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR