1,4-Dibrom-2,3-difluorbenzol 98%
|
|
BLDPBD54584-100GEA
454
EUR
BLDPBD54584-100G
BLDPBD54584-1G
BLDPBD54584-10G
BLDPBD54584-25G
BLDPBD54584-5G
BLDPBD54584-500G
1,4-Dibrom-2,3-difluorbenzol 98%
1,4-Dibromo-2,3-difluorobenzene
Storage condition: Keep in dark place, Sealed in dry, 2-8°C
Formel:
C₆H₂Br₂F₂ Molekulargewicht: 271,8849 g/mol Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD09835192 CAS-Nummer: 156682-52-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...