1-[(Ammoniooxy)methyl]-2-chlorbenzolchlorid 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD55296-1GEA 206 EUR
BLDPBD55296-1G BLDPBD55296-250MG BLDPBD55296-100MG
1-[(Ammoniooxy)methyl]-2-chlorbenzolchlorid 95%
1-[(Ammoniooxy)methyl]-2-chlorbenzolchlorid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₉Cl₂NO
Molekulargewicht: 194,0585 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01114592
CAS-Nummer: 5555-48-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR