3',5'-Dibrom-2'-hydroxyacetophenon 98%
|
|
BLDPBD5804-5GEA
59
EUR
BLDPBD5804-5G
BLDPBD5804-1G
BLDPBD5804-25G
BLDPBD5804-250MG
3',5'-Dibrom-2'-hydroxyacetophenon 98%
3',5'-Dibrom-2'-hydroxyacetophenon
Storage condition: Sealed in dry, Room Temperature
Formel:
C₈H₆Br₂O₂ Molekulargewicht: 293,95 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00191992 CAS-Nummer: 22362-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...