1-Methyl-5-indolboronsäurepinakolester 97%

BLDPBD59531-250MGEA 19 EUR
BLDPBD59531-250MG BLDPBD59531-5G BLDPBD59531-1G
1-Methyl-5-indolboronsäurepinakolester 97%
1-Methyl-5-indolboronsäurepinakolester

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₅H₂₀BNO₂
Molekulargewicht: 257,1358 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD05663865
CAS-Nummer: 837392-62-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR