3,5-Bis(trifluormethyl)phenylhydrazin 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD5969-250MGEA 18 EUR
BLDPBD5969-250MG
3,5-Bis(trifluormethyl)phenylhydrazin 98%
3,5-Bis(trifluormethyl)phenylhydrazin
Formel: C₈H₆F₆N₂
Molekulargewicht: 244,14 g/mol
MDL Number: MFCD00051606
CAS-Nummer: 886-35-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR