4-Cyanbenzamid 98%

BLDPBD5982-25GEA 350 EUR
BLDPBD5982-25G BLDPBD5982-1G BLDPBD5982-5G
4-Cyanbenzamid 98%
4-Cyanbenzamid

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₆N₂O
Molekulargewicht: 146,15 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00017133
CAS-Nummer: 3034-34-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR