4-(1-Pyrenyl)buttersäurehydrazid 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD60360-1GEA 1065 EUR
BLDPBD60360-1G BLDPBD60360-100MG BLDPBD60360-250MG
4-(1-Pyrenyl)buttersäurehydrazid 95%
4-(1-Pyrenyl)buttersäurehydrazid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₂₀H₁₈N₂O
Molekulargewicht: 302,3697 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00467260
CAS-Nummer: 55486-13-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR