1-Vinyl-2-pyrrolidon 98% stabilisiert


Stabilisierung: Stabilized with Ammonium hydroxide or NaOH 200 ppm

BLDPBD60560-25GEA 7 EUR
BLDPBD60560-25G
1-Vinyl-2-pyrrolidon 98% stabilisiert
1-Vinyl-2-pyrrolidon

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₉NO
Molekulargewicht: 111,1418 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00003197
CAS-Nummer: 88-12-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR