5'-Hydroxy-2'-methoxyacetophenon 97%

BLDPBD631795-250MGEA 41 EUR
BLDPBD631795-250MG BLDPBD631795-100MG BLDPBD631795-1G
5'-Hydroxy-2'-methoxyacetophenon 97%
5'-Hydroxy-2'-methoxyacetophenon

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₁₀O₃
Molekulargewicht: 166,1739 g/mol
Lagerungstemperatur: Raumtemperatur
CAS-Nummer: 31405-60-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR