2-(Diethylcarbamoyl)phenylboronsäure 95%

BLDPBD63341-100MGEA 39 EUR
BLDPBD63341-100MG BLDPBD63341-1G BLDPBD63341-250MG
2-(Diethylcarbamoyl)phenylboronsäure 95%
2-(Diethylcarbamoyl)phenylboronsäure

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₁H₁₆BNO₃
Molekulargewicht: 221,0600 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD01318988
CAS-Nummer: 129112-21-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR