2-Hydroxyethyl-phenylsulfid 97%

Lieferant: BLD PHARMATECH GMBH

BLDPBD63453-100GEA 105 EUR
BLDPBD63453-100G BLDPBD63453-25G BLDPBD63453-5G BLDPBD63453-10G
2-Hydroxyethyl-phenylsulfid 97%
2-Hydroxyethyl-phenylsulfid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₈H₁₀OS
Molekulargewicht: 154,23 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00002905
CAS-Nummer: 699-12-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR