4-Hydroxy-3-methylbenzaldehyd 95%

BLDPBD6383-100GEA 814 EUR
BLDPBD6383-100G BLDPBD6383-250MG BLDPBD6383-1G BLDPBD6383-10G BLDPBD6383-5G BLDPBD6383-25G
4-Hydroxy-3-methylbenzaldehyd 95%
4-Hydroxy-3-methylbenzaldehyde

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₈H₈O₂
Molekulargewicht: 136,1479 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00012360
CAS-Nummer: 15174-69-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR