1,4-Benzodioxan-6-boronsäure 96%
|
|
BLDPBD6445-1GEA
22
EUR
BLDPBD6445-1G
BLDPBD6445-250MG
BLDPBD6445-10G
BLDPBD6445-5G
BLDPBD6445-25G
BLDPBD6445-100G
1,4-Benzodioxan-6-boronsäure 96%
1,4-Benzodioxan-6-boronsäure
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formel:
C₈H₉BO₄ Molekulargewicht: 179,97 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01009696 CAS-Nummer: 164014-95-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...