2,4,6-T (2,4,6-Trichlorphenoxyessigsäure) 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD66225-5GEA 72 EUR
BLDPBD66225-5G BLDPBD66225-10G BLDPBD66225-1G BLDPBD66225-25G
2,4,6-T (2,4,6-Trichlorphenoxyessigsäure) 98%
2,4,6-T (2,4,6-Trichlorphenoxyessigsäure)

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₅Cl₃O₃
Molekulargewicht: 255,4825 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00053498
CAS-Nummer: 575-89-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR