Methyl-3-methoxysalicylat 97%
|
|
BLDPBD6719-10GEA
16
EUR
BLDPBD6719-10G
BLDPBD6719-5G
BLDPBD6719-100G
BLDPBD6719-25G
Methyl-3-methoxysalicylat 97%
Methyl-3-methoxysalicylat
Storage condition: Sealed in dry, Room Temperature
Formel:
C₉H₁₀O₄ Molekulargewicht: 182,1700 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00239511 CAS-Nummer: 6342-70-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...